The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030797.3 | NP_110424.1 | 4 - 5 | 446 | 79 |
XM_005262630.3 | XP_005262687.1 | 3 - 4 | 402 | 79 | |
XM_006712108.3 | XP_006712171.1 | 4 - 5 | 401 | 79 | |
GenBank mRNA | AK055334.1 | - | 4 - 5 | 414 | 79 |
AK315065.1 | - | 3 - 4 | 341 | 79 | |
AL136704.1 | - | 4 - 5 | 411 | 79 | |
BC038971.1 | - | 4 - 5 | 397 | 79 | |
CR533504.1 | - | 2 - 3 | 193 | 79 | |
DA188616.1 | - | 4 - 5 | 446 | 79 |