The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042618.1 | NP_001036083.1 | 9 - 10 | 930 | 110 |
NM_005484.3 | NP_005475.2 | 9 - 10 | 969 | 110 | |
XM_005267247.3 | XP_005267304.1 | 9 - 10 | 969 | 110 | |
XM_017020912.1 | XP_016876401.1 | 9 - 10 | 1503 | 110 | |
GenBank mRNA | AF085734.1 | - | 9 - 10 | 966 | 110 |
AJ236876.2 | - | 8 - 9 | 880 | 110 | |
AJ236912.1 | - | 9 - 10 | 904 | 110 | |
AK001980.1 | - | 9 - 10 | 946 | 110 | |
AK300971.1 | - | 7 - 8 | 687 | 110 |