The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270364.1 | NP_001257293.1 | 6 - 7 | 964 | 123 |
NM_001270365.1 | NP_001257294.1 | 4 - 5 | 763 | 123 | |
NM_012190.3 | NP_036322.2 | 6 - 7 | 1066 | 123 | |
NR_072979.1 | 6 - 7 | 1066 | 123 | ||
XM_006713481.2 | XP_006713544.1 | 6 - 7 | 805 | 123 | |
XM_011512355.1 | XP_011510657.1 | 6 - 7 | 798 | 123 | |
XM_017005613.1 | XP_016861102.1 | 6 - 7 | 1189 | 123 | |
XM_017005614.1 | XP_016861103.1 | 6 - 7 | 805 | 123 | |
GenBank mRNA | AB209299.1 | - | 6 - 7 | 873 | 123 |
AF052732.1 | - | 6 - 7 | 826 | 123 | |
AK222535.1 | - | 6 - 7 | 848 | 123 | |
AK222694.1 | - | 6 - 7 | 848 | 123 | |
AK294392.1 | - | 4 - 5 | 547 | 123 | |
AK294530.1 | - | 5 - 6 | 615 | 123 | |
AK307699.1 | - | 7 - 8 | 977 | 123 | |
AK308112.1 | - | 6 - 7 | 805 | 123 | |
AL133015.1 | - | 4 - 5 | 433 | 123 | |
BC027241.1 | - | 6 - 7 | 839 | 123 | |
CR749807.1 | - | 6 - 7 | 964 | 123 | |
KU178540.1 | - | 5 - 6 | 716 | 123 | |
KU178541.1 | - | 6 - 7 | 900 | 123 | |
KU178542.1 | - | 5 - 6 | 716 | 123 |