Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C12orf75
Entrez Gene ID
387882
Gene Name
chromosome 12 open reading frame 75
Gene Aliases
AGD3, OCC-1, OCC1
Chromosome Location
Chr.12: 105330636 - 105371518 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145199.1 NP_001138671.1 2 - 3 327 119
GenBank mRNA AJ311377.1 - 2 - 3 193 119
AJ311378.1 - 2 - 3 101 119
AK056999.1 - 2 - 3 199 119
BC013920.1 - 2 - 3 329 119
BC045812.1 - 2 - 3 291 119
BC061920.1 - 2 - 3 253 119
BC067897.1 - 2 - 3 301 119

View Full Product Details