The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282640.1 | NP_001269569.1 | 2 - 3 | 269 | 97 |
NM_001282643.1 | NP_001269572.1 | 2 - 3 | 269 | 97 | |
NM_022486.4 | NP_071931.2 | 2 - 3 | 269 | 97 | |
XM_011518915.1 | XP_011517217.1 | 2 - 3 | 492 | 97 | |
XM_011518916.2 | XP_011517218.1 | 2 - 3 | 492 | 97 | |
XM_011518917.1 | XP_011517219.1 | 2 - 3 | 492 | 97 | |
XM_011518918.1 | XP_011517220.1 | 2 - 3 | 492 | 97 | |
XM_017015037.1 | XP_016870526.1 | 2 - 3 | 492 | 97 | |
XM_017015038.1 | XP_016870527.1 | 2 - 3 | 492 | 97 | |
XM_017015039.1 | XP_016870528.1 | 2 - 3 | 492 | 97 | |
XM_017015040.1 | XP_016870529.1 | 2 - 3 | 492 | 97 | |
GenBank mRNA | AK056704.1 | - | 2 - 3 | 157 | 97 |
AK292868.1 | - | 2 - 3 | 269 | 97 | |
AY358746.1 | - | 2 - 3 | 248 | 97 | |
BC060770.1 | - | 2 - 3 | 239 | 97 | |
BX362255.2 | - | 2 - 3 | 256 | 97 | |
DB221801.1 | - | 2 - 3 | 270 | 97 |