Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUSD1
Entrez Gene ID
64420
Gene Name
sushi domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 112040781 - 112175520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282640.1 NP_001269569.1 3 - 4 430 69
NM_001282643.1 NP_001269572.1 3 - 4 430 69
NM_022486.4 NP_071931.2 3 - 4 430 69
XM_011518915.1 XP_011517217.1 3 - 4 653 69
XM_011518916.2 XP_011517218.1 3 - 4 653 69
XM_011518917.1 XP_011517219.1 3 - 4 653 69
XM_011518918.1 XP_011517220.1 3 - 4 653 69
XM_017015037.1 XP_016870526.1 3 - 4 653 69
XM_017015038.1 XP_016870527.1 3 - 4 653 69
XM_017015039.1 XP_016870528.1 3 - 4 653 69
XM_017015040.1 XP_016870529.1 3 - 4 653 69
GenBank mRNA AK292868.1 - 3 - 4 430 69
AY358746.1 - 3 - 4 409 69
BC060770.1 - 3 - 4 400 69
BX362255.2 - 3 - 4 417 69
DB221801.1 - 3 - 4 431 69

View Full Product Details