The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267040.1 | NP_001253969.1 | 10 - 11 | 2725 | 100 |
NM_004973.3 | NP_004964.2 | 10 - 11 | 3225 | 100 | |
XM_005249089.3 | XP_005249146.1 | 9 - 10 | 2563 | 100 | |
XM_011514579.2 | XP_011512881.1 | 12 - 13 | 3414 | 100 | |
XM_011514584.2 | XP_011512886.1 | 8 - 9 | 2479 | 100 | |
XM_017010833.1 | XP_016866322.1 | 12 - 13 | 3369 | 100 | |
XM_017010834.1 | XP_016866323.1 | 9 - 10 | 3083 | 100 | |
XM_017010835.1 | XP_016866324.1 | 8 - 9 | 2458 | 100 | |
GenBank mRNA | AK292861.1 | - | 10 - 11 | 3071 | 100 |
AK299349.1 | - | 9 - 10 | 2639 | 100 | |
AK303610.1 | - | 10 - 11 | 2668 | 100 | |
BC046246.1 | - | 10 - 11 | 2942 | 100 | |
U57592.1 | - | 10 - 11 | 2904 | 100 |