The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015980.4 | NP_057064.1 | 4 - 5 | 588 | 72 |
GenBank mRNA | AF113537.1 | - | 4 - 5 | 457 | 72 |
AF123767.1 | - | 4 - 5 | 457 | 72 | |
AF133422.1 | - | 4 - 5 | 441 | 72 | |
AK294383.1 | - | 3 - 4 | 371 | 72 | |
AK295206.1 | - | 3 - 4 | 271 | 72 | |
AK297196.1 | - | 4 - 5 | 394 | 72 | |
AK312359.1 | - | 4 - 5 | 475 | 72 | |
AK315949.1 | - | 2 - 3 | 203 | 72 | |
AK315952.1 | - | 3 - 4 | 285 | 72 | |
BC002619.2 | - | 4 - 5 | 471 | 72 | |
BC018873.2 | - | 4 - 5 | 464 | 72 | |
CR457142.1 | - | 3 - 4 | 326 | 72 |