Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB11FIP3
Entrez Gene ID
9727
Gene Name
RAB11 family interacting protein 3
Gene Aliases
CART1, Rab11-FIP3
Chromosome Location
Chr.16: 425621 - 522485 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142272.1 NP_001135744.1 7 - 8 939 73
NM_014700.3 NP_055515.1 9 - 10 1985 73
XM_005255713.3 XP_005255770.1 10 - 11 2155 73
XM_005255714.1 XP_005255771.1 8 - 9 1000 73
XM_005255715.4 XP_005255772.1 8 - 9 1806 73
XM_011522764.2 XP_011521066.1 8 - 9 1032 73
XM_011522765.2 XP_011521067.1 8 - 9 858 73
GenBank mRNA AB014565.1 - 9 - 10 2033 73
AF395731.1 - 9 - 10 2033 73
AK293644.1 - 7 - 8 854 73
AK303061.1 - 7 - 8 943 73
AY130007.1 - 1 - 2 54 73
BC051360.1 - 9 - 10 1984 73

View Full Product Details