The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142289.2 | NP_001135761.2 | 13 - 14 | 1552 | 83 |
NM_001142290.2 | NP_001135762.1 | 14 - 15 | 1618 | 83 | |
NM_001142291.2 | NP_001135763.2 | 13 - 14 | 1552 | 83 | |
NM_015246.3 | NP_056061.1 | 14 - 15 | 1618 | 83 | |
NR_102267.1 | 12 - 13 | 1442 | 83 | ||
XM_005255217.1 | XP_005255274.1 | 14 - 15 | 1621 | 83 | |
XM_005255218.1 | XP_005255275.1 | 13 - 14 | 1555 | 83 | |
XM_005255219.3 | XP_005255276.1 | 14 - 15 | 1621 | 83 | |
XM_005255220.3 | XP_005255277.1 | 14 - 15 | 1618 | 83 | |
XM_005255221.3 | XP_005255278.1 | 13 - 14 | 1552 | 83 | |
XM_011522441.1 | XP_011520743.1 | 14 - 15 | 1479 | 83 | |
XM_011522442.2 | XP_011520744.1 | 6 - 7 | 933 | 83 | |
XM_017023091.1 | XP_016878580.1 | 14 - 15 | 1621 | 83 | |
XM_017023092.1 | XP_016878581.1 | 13 - 14 | 1555 | 83 | |
XM_017023093.1 | XP_016878582.1 | 14 - 15 | 1476 | 83 | |
GenBank mRNA | AB011116.1 | - | 14 - 15 | 1574 | 83 |
AK299052.1 | - | 12 - 13 | 1424 | 83 | |
BC050389.1 | - | 14 - 15 | 1590 | 83 | |
EF471397.2 | - | 13 - 14 | 1415 | 83 | |
EF471398.2 | - | 13 - 14 | 1415 | 83 |