The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144924.1 | NP_001138396.1 | 5 - 6 | 742 | 59 |
NM_001199053.1 | NP_001185982.1 | 5 - 6 | 854 | 59 | |
NM_030941.2 | NP_112203.2 | 5 - 6 | 742 | 59 | |
XM_005255604.1 | XP_005255661.1 | 5 - 6 | 758 | 59 | |
XM_011545963.1 | XP_011544265.1 | 5 - 6 | 543 | 59 | |
XM_011545964.1 | XP_011544266.1 | 5 - 6 | 719 | 59 | |
XM_011545965.1 | XP_011544267.1 | 5 - 6 | 719 | 59 | |
XM_011545966.1 | XP_011544268.1 | 4 - 5 | 481 | 59 | |
XM_017023739.1 | XP_016879228.1 | 5 - 6 | 758 | 59 | |
GenBank mRNA | AF332193.1 | - | 5 - 6 | 676 | 59 |
AK314458.1 | - | 5 - 6 | 601 | 59 | |
AL136763.2 | - | 5 - 6 | 666 | 59 | |
BC007646.1 | - | 5 - 6 | 688 | 59 | |
BC068503.1 | - | 5 - 6 | 544 | 59 |