Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNNI2
Entrez Gene ID
7136
Gene Name
troponin I2, fast skeletal type
Gene Aliases
AMCD2B, DA2B, FSSV, fsTnI
Chromosome Location
Chr.11: 1839003 - 1841680 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145829.1 NP_001139301.1 6 - 7 340 68
NM_001145841.1 NP_001139313.1 4 - 5 301 68
NM_003282.3 NP_003273.1 6 - 7 332 68
GenBank mRNA AW984315.1 - 3 - 4 275 68
BC032148.2 - 6 - 7 340 68
BI833431.1 - 4 - 5 310 68
BP233165.1 - 6 - 7 354 68
BQ926240.1 - 6 - 7 341 68
L21715.1 - 5 - 6 305 68

View Full Product Details