The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134435.2 | NP_001127907.1 | 5 - 6 | 789 | 128 |
NM_001323027.1 | NP_001309956.1 | 5 - 6 | 1178 | 128 | |
NM_001323028.1 | NP_001309957.1 | 5 - 6 | 1001 | 128 | |
NM_001323029.1 | NP_001309958.1 | 6 - 7 | 1089 | 128 | |
NM_001323030.1 | NP_001309959.1 | 6 - 7 | 866 | 128 | |
NM_032263.4 | NP_115639.1 | 6 - 7 | 1068 | 128 | |
XM_017007316.1 | XP_016862805.1 | 6 - 7 | 1058 | 128 | |
XM_017007317.1 | XP_016862806.1 | 5 - 6 | 789 | 128 | |
GenBank mRNA | AK021729.1 | - | 2 - 3 | 613 | 128 |
AK027224.1 | - | 5 - 6 | 787 | 128 | |
AL136889.1 | - | 6 - 7 | 1052 | 128 | |
BC004816.1 | - | 3 - 4 | 430 | 128 | |
BC016156.1 | - | 6 - 7 | 1016 | 128 | |
BC136253.1 | - | 5 - 6 | 734 | 128 | |
CR533515.1 | - | 3 - 4 | 643 | 128 |