Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- IGF2
- Entrez Gene ID
-
3481
- Gene Name
- insulin like growth factor 2
- Gene Aliases
-
C11orf43, GRDF, IGF-II, PP9974
- Chromosome Location
-
Chr.11: 2129112 - 2149603 on Build GRCh38
- Gene Symbol
- INS-IGF2
- Entrez Gene ID
-
723961
- Gene Name
- INS-IGF2 readthrough
- Gene Aliases
-
INSIGF
- Chromosome Location
-
Chr.11: 2129112 - 2161209 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003512.3 |
|
5 - 6 |
877 |
81 |
GenBank mRNA |
AK291594.1 |
- |
2 - 3 |
|
81 |
DQ104205.1 |
- |
5 - 6 |
881 |
81 |
View Full Product Details