The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001408.2 | NP_001399.1 | 27 - 28 | 7869 | 70 |
XM_005270580.4 | XP_005270637.1 | 27 - 28 | 8183 | 70 | |
GenBank mRNA | AF234887.1 | - | 27 - 28 | 7870 | 70 |
AK124727.1 | - | 4 - 5 | 388 | 70 | |
D87469.2 | - | 27 - 28 | 7601 | 70 |