The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001408.2 | NP_001399.1 | 31 - 32 | 8397 | 117 |
XM_005270580.4 | XP_005270637.1 | 31 - 32 | 8711 | 117 | |
GenBank mRNA | AF234887.1 | - | 31 - 32 | 8398 | 117 |
AK124727.1 | - | 8 - 9 | 916 | 117 | |
AK127086.1 | - | 4 - 5 | 1236 | 117 | |
AK127745.1 | - | 6 - 7 | 2105 | 117 | |
D87469.2 | - | 31 - 32 | 8129 | 117 |