Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CELSR2
Entrez Gene ID
1952
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Gene Aliases
ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3
Chromosome Location
Chr.1: 109249705 - 109275756 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001408.2 NP_001399.1 33 - 34 8802 70
GenBank mRNA AF234887.1 - 33 - 34 8803 70
AK124727.1 - 10 - 11 1321 70
AK127745.1 - 8 - 9 2510 70
D87469.2 - 33 - 34 8534 70

View Full Product Details