The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001408.2 | NP_001399.1 | 9 - 10 | 5056 | 79 |
XM_005270580.4 | XP_005270637.1 | 9 - 10 | 5370 | 79 | |
GenBank mRNA | AF234887.1 | - | 9 - 10 | 5057 | 79 |
AK124727.1 | - | 1 - 2 | 66 | 79 | |
D87469.2 | - | 9 - 10 | 4788 | 79 |