The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024512.4 | NP_078788.2 | 2 - 3 | 525 | 90 |
XM_011534110.2 | XP_011532412.1 | 2 - 3 | 314 | 90 | |
XM_017007177.1 | XP_016862666.1 | 2 - 3 | 525 | 90 | |
GenBank mRNA | AJ308569.2 | - | 2 - 3 | 299 | 90 |
AK315636.1 | - | 2 - 3 | 308 | 90 | |
BC029118.1 | - | 2 - 3 | 297 | 90 | |
DA144445.1 | - | 2 - 3 | 525 | 90 | |
DA342323.1 | - | 2 - 3 | 496 | 90 |