Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A4
Entrez Gene ID
9122
Gene Name
solute carrier family 16 member 4
Gene Aliases
MCT4, MCT5
Chromosome Location
Chr.1: 110362851 - 110391970 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201546.1 NP_001188475.1 1 - 2 219 77
NM_001201547.1 NP_001188476.1 1 - 2 219 77
NM_001201548.1 NP_001188477.1 1 - 2 219 77
NM_001201549.1 NP_001188478.1 1 - 2 219 77
NM_001319220.1 NP_001306149.1 1 - 2 219 77
NM_004696.2 NP_004687.1 1 - 2 219 77
XM_006711033.3 XP_006711096.1 1 - 2 1105 77
GenBank mRNA AK129985.1 - 1 - 2 163 77
AK223307.1 - 1 - 2 163 77
AK290720.1 - 1 - 2 163 77
AK295946.1 - 1 - 2 185 77
AK298539.1 - 1 - 2 163 77
AK313735.1 - 1 - 2 181 77
AL833619.1 - 1 - 2 160 77
BC021664.1 - 1 - 2 189 77
BE893159.1 - 1 - 2 60 77
BP345760.1 - 1 - 2 219 77
BX647948.1 - 1 - 2 185 77
U59185.1 - 1 - 2 151 77

View Full Product Details