The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201547.1 | NP_001188476.1 | 3 - 4 | 479 | 80 |
NM_001201549.1 | NP_001188478.1 | 4 - 5 | 612 | 80 | |
NM_001319220.1 | NP_001306149.1 | 3 - 4 | 479 | 80 | |
NM_004696.2 | NP_004687.1 | 4 - 5 | 612 | 80 | |
XM_005271317.4 | XP_005271374.1 | 5 - 6 | 1630 | 80 | |
GenBank mRNA | AK129985.1 | - | 4 - 5 | 556 | 80 |
AK223307.1 | - | 4 - 5 | 556 | 80 | |
AK295946.1 | - | 3 - 4 | 445 | 80 | |
AK313735.1 | - | 4 - 5 | 574 | 80 | |
BC021664.1 | - | 4 - 5 | 582 | 80 | |
BE893159.1 | - | 3 - 4 | 320 | 80 | |
BX647948.1 | - | 3 - 4 | 445 | 80 | |
U59185.1 | - | 4 - 5 | 544 | 80 |