The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201549.1 | NP_001188478.1 | 5 - 6 | 778 | 95 |
NM_001319220.1 | NP_001306149.1 | 4 - 5 | 645 | 95 | |
GenBank mRNA | BC021664.1 | - | 5 - 6 | 748 | 95 |
BX647948.1 | - | 4 - 5 | 611 | 95 |