The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001971.5 | NP_001962.3 | 4 - 5 | 364 | 105 |
GenBank mRNA | AF120493.1 | - | 4 - 5 | 364 | 105 |
BC069454.1 | - | 4 - 5 | 364 | 105 | |
BC075091.2 | - | 4 - 5 | 345 | 105 |