The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170738.1 | NP_001164209.1 | 13 - 14 | 3111 | 88 |
XM_011520961.1 | XP_011519263.1 | 12 - 13 | 2614 | 88 | |
GenBank mRNA | AB029033.1 | - | 10 - 11 | 1786 | 88 |