The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170738.1 | NP_001164209.1 | 2 - 3 | 624 | 109 |
XM_011520958.2 | XP_011519260.1 | 2 - 3 | 712 | 109 | |
XM_011520959.2 | XP_011519261.1 | 2 - 3 | 712 | 109 | |
XM_011520960.1 | XP_011519262.1 | 2 - 3 | 711 | 109 | |
XM_017019310.1 | XP_016874799.1 | 2 - 3 | 712 | 109 | |
XM_017019311.1 | XP_016874800.1 | 2 - 3 | 712 | 109 | |
GenBank mRNA | BC042067.1 | - | 2 - 3 | 470 | 109 |