Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JAK3
Entrez Gene ID
3718
Gene Name
Janus kinase 3
Gene Aliases
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
Chromosome Location
Chr.19: 17824782 - 17848071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000215.3 NP_000206.2 15 - 16 2152 106
XM_005259896.3 XP_005259953.1 15 - 16 2191 106
XM_011527990.1 XP_011526292.1 15 - 16 2191 106
GenBank mRNA U09607.1 - 14 - 15 2147 106
U31601.1 - 15 - 16 2147 106

View Full Product Details