Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JAK3
Entrez Gene ID
3718
Gene Name
Janus kinase 3
Gene Aliases
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
Chromosome Location
Chr.19: 17824782 - 17848071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000215.3 NP_000206.2 21 - 22 3076 86
XM_005259896.3 XP_005259953.1 21 - 22 3115 86
GenBank mRNA U08340.1 - 1 - 2 132 86
U09607.1 - 20 - 21 3071 86
U31601.1 - 21 - 22 3071 86

View Full Product Details