Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JAK3
Entrez Gene ID
3718
Gene Name
Janus kinase 3
Gene Aliases
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
Chromosome Location
Chr.19: 17824782 - 17848071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000215.3 NP_000206.2 7 - 8 1086 115
XM_005259896.3 XP_005259953.1 7 - 8 1125 115
XM_011527990.1 XP_011526292.1 7 - 8 1125 115
XM_011527991.2 XP_011526293.1 7 - 8 1125 115
GenBank mRNA AK296383.1 - 7 - 8 1087 115
BC028068.1 - 7 - 8 1076 115
KU178153.1 - 6 - 7 986 115
KU178154.1 - 6 - 7 986 115
U09607.1 - 6 - 7 1081 115
U31601.1 - 7 - 8 1081 115

View Full Product Details