The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099856.4 | NP_001093326.2 | 9 - 10 | 1344 | 150 |
NM_001099857.2 | NP_001093327.1 | 9 - 10 | 1530 | 150 | |
NM_001145255.2 | NP_001138727.1 | 7 - 8 | 1233 | 150 | |
NM_001321396.1 | NP_001308325.1 | 9 - 10 | 1279 | 150 | |
NM_001321397.1 | NP_001308326.1 | 9 - 10 | 1527 | 150 | |
NM_003639.4 | NP_003630.1 | 9 - 10 | 1374 | 150 | |
GenBank mRNA | AF062089.1 | - | 9 - 10 | 1226 | 150 |
AF074382.1 | - | 9 - 10 | 1264 | 150 | |
AF091453.1 | - | 9 - 10 | 1301 | 150 | |
AF261086.1 | - | 9 - 10 | 1375 | 150 | |
AK000593.1 | - | 7 - 8 | 938 | 150 | |
AY114157.1 | - | 9 - 10 | 1340 | 150 | |
BC000299.2 | - | 9 - 10 | 1236 | 150 | |
BC012114.1 | - | 9 - 10 | 1340 | 150 | |
BC046922.1 | - | 9 - 10 | 1231 | 150 | |
BC050612.2 | - | 9 - 10 | 1263 | 150 | |
BT019621.1 | - | 8 - 9 | 1116 | 150 | |
CA916723.1 | - | 9 - 10 | 1238 | 150 |