The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017789.4 | NP_060259.4 | 11 - 12 | 1492 | 69 |
XM_006712606.3 | XP_006712669.2 | 11 - 12 | 1596 | 69 | |
XM_011511378.2 | XP_011509680.1 | 11 - 12 | 1600 | 69 | |
XM_011511379.2 | XP_011509681.1 | 11 - 12 | 1597 | 69 | |
XM_011511380.1 | XP_011509682.1 | 11 - 12 | 1935 | 69 | |
XM_011511381.1 | XP_011509683.1 | 11 - 12 | 1569 | 69 | |
XM_011511382.2 | XP_011509684.1 | 11 - 12 | 1528 | 69 | |
XM_011511383.1 | XP_011509685.1 | 11 - 12 | 1540 | 69 | |
XM_017004393.1 | XP_016859882.1 | 10 - 11 | 2022 | 69 | |
XM_017004394.1 | XP_016859883.1 | 9 - 10 | 1144 | 69 | |
GenBank mRNA | AB051526.1 | - | 9 - 10 | 1722 | 69 |
AF258577.1 | - | 1 - 2 | 50 | 69 | |
AY358842.1 | - | 11 - 12 | 1465 | 69 | |
BC017476.1 | - | 5 - 6 | 768 | 69 | |
BC062984.1 | - | 3 - 4 | 464 | 69 | |
BC109103.1 | - | 11 - 12 | 1421 | 69 | |
BC109104.1 | - | 11 - 12 | 1418 | 69 |