The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017789.4 | NP_060259.4 | 4 - 5 | 484 | 62 |
XM_006712606.3 | XP_006712669.2 | 4 - 5 | 588 | 62 | |
XM_011511378.2 | XP_011509680.1 | 4 - 5 | 592 | 62 | |
XM_011511379.2 | XP_011509681.1 | 4 - 5 | 589 | 62 | |
XM_011511380.1 | XP_011509682.1 | 4 - 5 | 927 | 62 | |
XM_011511381.1 | XP_011509683.1 | 4 - 5 | 561 | 62 | |
XM_011511382.2 | XP_011509684.1 | 4 - 5 | 520 | 62 | |
XM_011511383.1 | XP_011509685.1 | 4 - 5 | 532 | 62 | |
XM_017004393.1 | XP_016859882.1 | 3 - 4 | 1014 | 62 | |
XM_017004394.1 | XP_016859883.1 | 2 - 3 | 136 | 62 | |
GenBank mRNA | AB051526.1 | - | 2 - 3 | 714 | 62 |
AY358842.1 | - | 4 - 5 | 457 | 62 | |
BC109103.1 | - | 4 - 5 | 413 | 62 | |
BC109104.1 | - | 4 - 5 | 410 | 62 | |
BI518922.1 | - | 4 - 5 | 485 | 62 |