The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144994.7 | NP_659431.5 | 6 - 7 | 655 | 106 |
GenBank mRNA | AF492401.1 | - | 6 - 7 | 631 | 106 |
AJ315766.1 | - | 5 - 6 | 599 | 106 | |
AK057011.1 | - | 6 - 7 | 655 | 106 | |
AK092564.1 | - | 5 - 6 | 529 | 106 | |
AY196212.1 | - | 6 - 7 | 639 | 106 | |
BC107056.2 | - | 6 - 7 | 650 | 106 |