Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTERF3
Entrez Gene ID
51001
Gene Name
mitochondrial transcription termination factor 3
Gene Aliases
CGI-12, MTERFD1
Chromosome Location
Chr.8: 96239398 - 96261613 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286643.1 NP_001273572.1 1 - 2 115 94
NM_015942.4 NP_057026.3 1 - 2 115 94
XM_006716570.3 XP_006716633.1 1 - 2 108 94
GenBank mRNA AK309002.1 - 1 - 2 112 94
BC012995.2 - 1 - 2 70 94
HY116693.1 - 1 - 2 115 94

View Full Product Details