The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008697.1 | NP_001008697.1 | 15 - 16 | 2485 | 124 |
NM_012143.2 | NP_036275.1 | 14 - 15 | 2408 | 124 | |
XM_011530079.1 | XP_011528381.1 | 15 - 16 | 2468 | 124 | |
XM_017028735.1 | XP_016884224.1 | 15 - 16 | 2626 | 124 | |
GenBank mRNA | AF070662.1 | - | 1 - 2 | 481 | 124 |
AK025443.1 | - | 13 - 14 | 2532 | 124 | |
AK025739.1 | - | 10 - 11 | 1866 | 124 | |
AK298303.1 | - | 10 - 11 | 1468 | 124 | |
AK299964.1 | - | 9 - 10 | 1234 | 124 | |
AL050258.1 | - | 14 - 15 | 2421 | 124 | |
AL080147.1 | - | 14 - 15 | 2388 | 124 | |
BC011599.2 | - | 14 - 15 | 2385 | 124 | |
BC033080.1 | - | 14 - 15 | 2386 | 124 | |
BT007274.1 | - | 11 - 12 | 2158 | 124 | |
CT841511.2 | - | 12 - 13 | 2260 | 124 |