The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005015.3 | NP_005006.3 | 2 - 3 | 403 | 76 |
GenBank mRNA | AK294849.1 | - | 2 - 3 | 250 | 76 |
AK298249.1 | - | 2 - 3 | 247 | 76 | |
BC001669.2 | - | 2 - 3 | 229 | 76 | |
BC112331.1 | - | 2 - 3 | 403 | 76 | |
BP303816.1 | - | 2 - 3 | 235 | 76 | |
BP330984.1 | - | 2 - 3 | 267 | 76 | |
BX248001.1 | - | 2 - 3 | 460 | 76 | |
BX248295.1 | - | 2 - 3 | 222 | 76 | |
X80695.1 | - | 2 - 3 | 229 | 76 |