Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THBS4
Entrez Gene ID
7060
Gene Name
thrombospondin 4
Gene Aliases
TSP-4, TSP4
Chromosome Location
Chr.5: 79991296 - 80083287 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306212.1 NP_001293141.1 14 - 15 1785 66
NM_001306213.1 NP_001293142.1 14 - 15 1837 66
NM_001306214.1 NP_001293143.1 13 - 14 1779 66
NM_003248.5 NP_003239.2 13 - 14 1951 66
XM_017009798.1 XP_016865287.1 13 - 14 1931 66
XM_017009799.1 XP_016865288.1 13 - 14 1931 66
XM_017009800.1 XP_016865289.1 13 - 14 1931 66
GenBank mRNA AK302842.1 - 13 - 14 1779 66
AK313587.1 - 13 - 14 1767 66
BC050456.1 - 14 - 15 2035 66
Z19585.1 - 13 - 14 1739 66

View Full Product Details