Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THBS4
Entrez Gene ID
7060
Gene Name
thrombospondin 4
Gene Aliases
TSP-4, TSP4
Chromosome Location
Chr.5: 79991296 - 80083287 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306212.1 NP_001293141.1 20 - 21 2585 58
NM_001306213.1 NP_001293142.1 20 - 21 2637 58
NM_001306214.1 NP_001293143.1 19 - 20 2579 58
NM_003248.5 NP_003239.2 19 - 20 2751 58
GenBank mRNA AK302842.1 - 19 - 20 2579 58
AK313587.1 - 19 - 20 2567 58
BC050456.1 - 20 - 21 2835 58
Z19585.1 - 19 - 20 2539 58

View Full Product Details