Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THBS4
Entrez Gene ID
7060
Gene Name
thrombospondin 4
Gene Aliases
TSP-4, TSP4
Chromosome Location
Chr.5: 79991296 - 80083287 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306212.1 NP_001293141.1 5 - 6 721 69
NM_001306213.1 NP_001293142.1 5 - 6 773 69
NM_001306214.1 NP_001293143.1 4 - 5 715 69
NM_003248.5 NP_003239.2 4 - 5 887 69
XM_017009798.1 XP_016865287.1 4 - 5 867 69
XM_017009799.1 XP_016865288.1 4 - 5 867 69
XM_017009800.1 XP_016865289.1 4 - 5 867 69
GenBank mRNA AK302842.1 - 4 - 5 715 69
AK313587.1 - 4 - 5 703 69
BC050456.1 - 5 - 6 971 69
Z19585.1 - 4 - 5 675 69

View Full Product Details