The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256041.1 | NP_001242970.1 | 23 - 24 | 2639 | 66 |
NM_005379.3 | NP_005370.1 | 22 - 23 | 2602 | 66 | |
XM_011538373.2 | XP_011536675.1 | 22 - 23 | 2570 | 66 | |
GenBank mRNA | AF009961.1 | - | 21 - 22 | 2351 | 66 |
AF105424.1 | - | 22 - 23 | 2560 | 66 | |
AF127026.1 | - | 22 - 23 | 2595 | 66 | |
AK300381.1 | - | 18 - 19 | 2183 | 66 | |
AK312431.1 | - | 23 - 24 | 2639 | 66 | |
BC059387.1 | - | 22 - 23 | 2602 | 66 |