Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNX5
Entrez Gene ID
27131
Gene Name
sorting nexin 5
Gene Aliases
-
Chromosome Location
Chr.20: 17941596 - 17968991 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282454.1 NP_001269383.1 9 - 10 1073 133
NM_014426.3 NP_055241.1 9 - 10 1201 133
NM_152227.2 NP_689413.1 10 - 11 1156 133
GenBank mRNA AF121855.1 - 10 - 11 1012 133
AK001793.1 - 9 - 10 918 133
AK026227.1 - 2 - 3 113 133
AK054634.1 - 10 - 11 1944 133
AK129505.1 - 9 - 10 918 133
AK223308.1 - 9 - 10 886 133
AK296947.1 - 10 - 11 1025 133
BC062638.1 - 9 - 10 858 133
BC093623.1 - 9 - 10 1127 133
BC093980.1 - 9 - 10 1127 133
BC143274.1 - 9 - 10 1127 133

View Full Product Details