The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022131.2 | NP_071414.2 | 1 - 2 | 293 | 111 |
GenBank mRNA | AJ278018.1 | - | 1 - 2 | 113 | 111 |
AK096432.1 | - | 1 - 2 | 293 | 111 | |
AK315308.1 | - | 1 - 2 | 293 | 111 | |
AY753303.1 | - | 1 - 2 | 103 | 111 | |
BC103496.1 | - | 1 - 2 | 113 | 111 | |
BC103506.1 | - | 1 - 2 | 112 | 111 | |
BC104485.1 | - | 1 - 2 | 110 | 111 |