The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018241.2 | NP_060711.2 | 8 - 9 | 1448 | 75 |
GenBank mRNA | AF305823.1 | - | 1 - 2 | 124 | 75 |
AK000169.1 | - | 5 - 6 | 601 | 75 | |
AK001708.1 | - | 8 - 9 | 1391 | 75 | |
BM712423.1 | - | 2 - 3 | 168 | 75 |