Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCE1
Entrez Gene ID
6059
Gene Name
ATP binding cassette subfamily E member 1
Gene Aliases
ABC38, OABP, RLI, RLI1, RNASEL1, RNASELI, RNS4I
Chromosome Location
Chr.4: 145098004 - 145129524 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040876.1 NP_001035809.1 1 - 2 414 91

View Full Product Details