Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO7
Entrez Gene ID
4008
Gene Name
LIM domain 7
Gene Aliases
FBX20, FBXO20, LOMP
Chromosome Location
Chr.13: 75620434 - 75859870 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306080.1 NP_001293009.1 24 - 25 4402 96
NM_005358.5 NP_005349.3 23 - 24 4446 96
NM_015842.2 NP_056667.2 21 - 22 4093 96
XM_011535075.2 XP_011533377.1 25 - 26 4290 96
XM_011535078.1 XP_011533380.1 24 - 25 4248 96
XM_011535080.2 XP_011533382.1 25 - 26 4209 96
XM_011535082.1 XP_011533384.1 24 - 25 4167 96
XM_011535086.1 XP_011533388.1 23 - 24 3963 96
XM_011535094.1 XP_011533396.1 18 - 19 2871 96
XM_011535095.1 XP_011533397.1 21 - 22 3042 96
XM_017020574.1 XP_016876063.1 25 - 26 4290 96
XM_017020575.1 XP_016876064.1 24 - 25 4248 96
XM_017020576.1 XP_016876065.1 24 - 25 4233 96
XM_017020577.1 XP_016876066.1 25 - 26 4209 96
XM_017020578.1 XP_016876067.1 23 - 24 4191 96
XM_017020579.1 XP_016876068.1 24 - 25 4167 96
XM_017020580.1 XP_016876069.1 24 - 25 4086 96
XM_017020581.1 XP_016876070.1 24 - 25 4212 96
XM_017020582.1 XP_016876071.1 24 - 25 4005 96
XM_017020583.1 XP_016876072.1 24 - 25 4005 96
XM_017020584.1 XP_016876073.1 23 - 24 4008 96
XM_017020585.1 XP_016876074.1 23 - 24 3804 96
XM_017020586.1 XP_016876075.1 23 - 24 3723 96
XM_017020587.1 XP_016876076.1 22 - 23 3681 96
XM_017020588.1 XP_016876077.1 22 - 23 3762 96
XM_017020589.1 XP_016876078.1 22 - 23 3885 96
XM_017020590.1 XP_016876079.1 22 - 23 3681 96
XM_017020591.1 XP_016876080.1 24 - 25 4373 96
XM_017020592.1 XP_016876081.1 24 - 25 4256 96
XM_017020593.1 XP_016876082.1 23 - 24 4169 96
XM_017020594.1 XP_016876083.1 23 - 24 4052 96
XM_017020595.1 XP_016876084.1 22 - 23 4046 96
XM_017020596.1 XP_016876085.1 22 - 23 3084 96
XM_017020597.1 XP_016876086.1 21 - 22 3054 96
XM_017020598.1 XP_016876087.1 22 - 23 4046 96
XM_017020599.1 XP_016876088.1 22 - 23 3887 96
XM_017020600.1 XP_016876089.1 22 - 23 3003 96
XM_017020601.1 XP_016876090.1 21 - 22 3764 96
XM_017020602.1 XP_016876091.1 21 - 22 3647 96
XM_017020603.1 XP_016876092.1 21 - 22 3764 96
XM_017020604.1 XP_016876093.1 21 - 22 3167 96
GenBank mRNA AB020665.2 - 21 - 22 4023 96
AF144237.1 - 12 - 13 1906 96
AF330045.1 - 23 - 24 4445 96
AK055465.1 - 16 - 17 3058 96
AK074265.1 - 8 - 9 895 96
AK092052.1 - 22 - 23 3365 96
AK304051.1 - 19 - 20 2984 96
FJ711162.1 - 24 - 25 4032 96
U90654.1 - 1 - 2 162 96

View Full Product Details