Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO7
Entrez Gene ID
4008
Gene Name
LIM domain 7
Gene Aliases
FBX20, FBXO20, LOMP
Chromosome Location
Chr.13: 75620434 - 75859870 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306080.1 NP_001293009.1 3 - 4 576 88
NM_005358.5 NP_005349.3 4 - 5 1622 88
XM_011535075.2 XP_011533377.1 2 - 3 137 88
XM_011535078.1 XP_011533380.1 2 - 3 137 88
XM_011535080.2 XP_011533382.1 2 - 3 137 88
XM_011535082.1 XP_011533384.1 2 - 3 137 88
XM_011535086.1 XP_011533388.1 2 - 3 137 88
XM_011535095.1 XP_011533397.1 2 - 3 137 88
XM_017020574.1 XP_016876063.1 2 - 3 137 88
XM_017020575.1 XP_016876064.1 2 - 3 137 88
XM_017020576.1 XP_016876065.1 2 - 3 137 88
XM_017020577.1 XP_016876066.1 2 - 3 137 88
XM_017020578.1 XP_016876067.1 2 - 3 137 88
XM_017020579.1 XP_016876068.1 2 - 3 137 88
XM_017020580.1 XP_016876069.1 2 - 3 137 88
XM_017020582.1 XP_016876071.1 2 - 3 137 88
XM_017020583.1 XP_016876072.1 2 - 3 137 88
XM_017020585.1 XP_016876074.1 2 - 3 137 88
XM_017020586.1 XP_016876075.1 2 - 3 137 88
XM_017020587.1 XP_016876076.1 2 - 3 137 88
XM_017020588.1 XP_016876077.1 2 - 3 137 88
XM_017020590.1 XP_016876079.1 2 - 3 137 88
XM_017020596.1 XP_016876085.1 2 - 3 137 88
XM_017020597.1 XP_016876086.1 2 - 3 137 88
XM_017020600.1 XP_016876089.1 2 - 3 137 88
GenBank mRNA AF330045.1 - 4 - 5 1621 88
AK092052.1 - 3 - 4 541 88
FJ711162.1 - 3 - 4 206 88

View Full Product Details