Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO7
Entrez Gene ID
4008
Gene Name
LIM domain 7
Gene Aliases
FBX20, FBXO20, LOMP
Chromosome Location
Chr.13: 75620434 - 75859870 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306080.1 NP_001293009.1 30 - 31 5247 70
NM_005358.5 NP_005349.3 29 - 30 5291 70
NM_015842.2 NP_056667.2 26 - 27 4829 70
XM_011535075.2 XP_011533377.1 31 - 32 5135 70
XM_011535078.1 XP_011533380.1 30 - 31 5093 70
XM_011535080.2 XP_011533382.1 31 - 32 5054 70
XM_011535082.1 XP_011533384.1 30 - 31 5012 70
XM_011535086.1 XP_011533388.1 28 - 29 4699 70
XM_011535094.1 XP_011533396.1 24 - 25 3716 70
XM_011535095.1 XP_011533397.1 27 - 28 3887 70
XM_017020583.1 XP_016876072.1 30 - 31 4850 70
XM_017020588.1 XP_016876077.1 27 - 28 4498 70
XM_017020589.1 XP_016876078.1 27 - 28 4621 70
XM_017020590.1 XP_016876079.1 27 - 28 4417 70
XM_017020595.1 XP_016876084.1 28 - 29 4891 70
XM_017020598.1 XP_016876087.1 27 - 28 4782 70
XM_017020600.1 XP_016876089.1 28 - 29 3848 70
XM_017020601.1 XP_016876090.1 27 - 28 4609 70
XM_017020602.1 XP_016876091.1 27 - 28 4492 70
XM_017020603.1 XP_016876092.1 26 - 27 4500 70
GenBank mRNA AB020665.2 - 26 - 27 4759 70
AF144237.1 - 18 - 19 2751 70
AF330045.1 - 29 - 30 5290 70
AK304051.1 - 25 - 26 3829 70
U90654.1 - 7 - 8 1007 70

View Full Product Details