Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMO7
Entrez Gene ID
4008
Gene Name
LIM domain 7
Gene Aliases
FBX20, FBXO20, LOMP
Chromosome Location
Chr.13: 75620434 - 75859870 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306080.1 NP_001293009.1 10 - 11 2287 78
NM_015842.2 NP_056667.2 7 - 8 1978 78
XM_011535075.2 XP_011533377.1 10 - 11 2052 78
XM_011535078.1 XP_011533380.1 10 - 11 2052 78
XM_011535080.2 XP_011533382.1 10 - 11 2052 78
XM_011535082.1 XP_011533384.1 10 - 11 2052 78
XM_011535086.1 XP_011533388.1 9 - 10 1848 78
XM_017020574.1 XP_016876063.1 10 - 11 2052 78
XM_017020575.1 XP_016876064.1 10 - 11 2052 78
XM_017020576.1 XP_016876065.1 10 - 11 2052 78
XM_017020577.1 XP_016876066.1 10 - 11 2052 78
XM_017020578.1 XP_016876067.1 10 - 11 2052 78
XM_017020579.1 XP_016876068.1 10 - 11 2052 78
XM_017020580.1 XP_016876069.1 9 - 10 1848 78
XM_017020581.1 XP_016876070.1 9 - 10 1974 78
XM_017020582.1 XP_016876071.1 9 - 10 1848 78
XM_017020583.1 XP_016876072.1 9 - 10 1848 78
XM_017020584.1 XP_016876073.1 8 - 9 1770 78
XM_017020585.1 XP_016876074.1 8 - 9 1566 78
XM_017020586.1 XP_016876075.1 8 - 9 1566 78
XM_017020587.1 XP_016876076.1 8 - 9 1566 78
XM_017020588.1 XP_016876077.1 8 - 9 1566 78
XM_017020589.1 XP_016876078.1 8 - 9 1770 78
XM_017020590.1 XP_016876079.1 8 - 9 1566 78
XM_017020591.1 XP_016876080.1 9 - 10 2135 78
XM_017020592.1 XP_016876081.1 9 - 10 2018 78
XM_017020593.1 XP_016876082.1 8 - 9 1931 78
XM_017020594.1 XP_016876083.1 8 - 9 1814 78
XM_017020595.1 XP_016876084.1 8 - 9 1931 78
XM_017020598.1 XP_016876087.1 8 - 9 1931 78
XM_017020599.1 XP_016876088.1 7 - 8 1649 78
XM_017020601.1 XP_016876090.1 7 - 8 1649 78
XM_017020602.1 XP_016876091.1 7 - 8 1532 78
XM_017020603.1 XP_016876092.1 7 - 8 1649 78
GenBank mRNA AB020665.2 - 7 - 8 1908 78
AK055465.1 - 2 - 3 943 78
FJ711162.1 - 10 - 11 1917 78

View Full Product Details