The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145847.1 | NP_001139319.1 | 3 - 4 | 784 | 117 |
NM_001145848.1 | NP_001139320.1 | 3 - 4 | 500 | 117 | |
NM_001145851.1 | NP_001139323.1 | 2 - 3 | 477 | 117 | |
NM_001145852.1 | NP_001139324.1 | 2 - 3 | 477 | 117 | |
XM_005248195.4 | XP_005248252.1 | 3 - 4 | 600 | 117 | |
XM_005248196.4 | XP_005248253.1 | 3 - 4 | 600 | 117 | |
XM_011513899.2 | XP_011512201.2 | 3 - 4 | 600 | 117 | |
XM_017008799.1 | XP_016864288.1 | 3 - 4 | 630 | 117 | |
XM_017008803.1 | XP_016864292.1 | 3 - 4 | 575 | 117 | |
XM_017008804.1 | XP_016864293.1 | 3 - 4 | 645 | 117 | |
XM_017008805.1 | XP_016864294.1 | 3 - 4 | 518 | 117 | |
GenBank mRNA | AF117225.1 | - | 3 - 4 | 545 | 117 |
AF507034.1 | - | 2 - 3 | 302 | 117 | |
AK027420.1 | - | 3 - 4 | 523 | 117 | |
AK027422.1 | - | 3 - 4 | 545 | 117 | |
AK075411.1 | - | 3 - 4 | 500 | 117 | |
AK226033.1 | - | 3 - 4 | 523 | 117 | |
AK314529.1 | - | 3 - 4 | 545 | 117 | |
AY449689.1 | - | 2 - 3 | 265 | 117 | |
AY449690.1 | - | 2 - 3 | 265 | 117 | |
AY449691.1 | - | 2 - 3 | 265 | 117 | |
BC012089.1 | - | 3 - 4 | 500 | 117 | |
HQ628626.1 | - | 2 - 3 | 265 | 117 |