Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL24A1
Entrez Gene ID
255631
Gene Name
collagen type XXIV alpha 1
Gene Aliases
-
Chromosome Location
Chr.1: 85728507 - 86156987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152890.5 NP_690850.2 59 - 60 5037 97
XM_017000924.1 XP_016856413.1 61 - 62 5210 97
XM_017000925.1 XP_016856414.1 61 - 62 5165 97
XM_017000926.1 XP_016856415.1 60 - 61 5368 97
XM_017000927.1 XP_016856416.1 59 - 60 5353 97
XM_017000928.1 XP_016856417.1 42 - 43 4119 97
XM_017000929.1 XP_016856418.1 41 - 42 4073 97
GenBank mRNA AB209686.1 - 34 - 35 4366 97
AY244357.1 - 59 - 60 5027 97
BC113654.1 - 59 - 60 5034 97
BC117170.1 - 59 - 60 5034 97
BC143715.1 - 59 - 60 5006 97
BC143717.1 - 59 - 60 5030 97
BC143718.1 - 58 - 59 5007 97

View Full Product Details