The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021977.3 | NP_068812.1 | 10 - 11 | 1657 | 73 |
XM_005267106.4 | XP_005267163.1 | 10 - 11 | 1384 | 73 | |
XM_011536075.2 | XP_011534377.1 | 10 - 11 | 8065 | 73 | |
XM_011536076.2 | XP_011534378.1 | 10 - 11 | 1511 | 73 | |
XM_017011203.1 | XP_016866692.1 | 10 - 11 | 1285 | 73 | |
GenBank mRNA | AF078749.1 | - | 1 - 2 | 75 | 73 |
AJ001417.1 | - | 10 - 11 | 1637 | 73 | |
AK314590.1 | - | 10 - 11 | 1652 | 73 | |
BC144280.1 | - | 9 - 10 | 1598 | 73 |